Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
BT Lab Systems Digital Magnetic Stirrer with Timer, G-Biosciences
Supplier: G-Biosciences
BT1005
10858-500EA 714.13 USD InStock
10858-500
BT Lab Systems Digital Magnetic Stirrer with Timer, G-Biosciences
Stirrers Magnetic Stirrers
A digital magnetic stirrer designed for stirring <3L containers at a maximum speed of 1,800rpm.
  • DC brushless motor for stable operation, low noise, long life and no risk of sparks
  • Stainless steel shell, firm and durable, thermo stability and anti-corrosion
  • Separate digital display for speed and timer
Digital timer included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty