Would you like to log in again?
BT Lab Systems Microplate Incubators, G-Biosciences
Supplier: G-Biosciences
BT1102 BT1101
10858-508EA 1076.76 USD InStock
10858-508 10858-510
BT Lab Systems Microplate Incubators, G-Biosciences
Incubators Microplate Incubators
A microplate incubator ideal for solution incubation of ELISA plates (96-/384-well) and cell culture plates (24-/48-/96-well).
  • LCD displays set and actual temperature and time simultaneously
  • User friendly touch operation interface
  • Sleak design offers small footprint and easy cleaning
  • Heats microplates from top and bottom to ensure uniformity
  • Alarm signal after operation end
  • Option of 2 plate or 4 plate capacity
A microplate incubator designed for heating plates up to 70°C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty