Would you like to log in again?
Acid Encapsulating & Neutralizing Pillow, PIG®
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIL352
10860-080BX 158.87 USD InStock
10860-080
Acid Encapsulating & Neutralizing Pillow, PIG®
Spill Control Materials
Encapsulate and neutralize the most common acid spills with the Acid Encapsulating & Neutralizing Pillow.
  • Great for battery charging
  • Identify best times for handling
  • Instantly absorbs up to 98% sulfuric acid conectration
Encapsulate and neutralize the most common acid spills; great for battery charging, transport and storage areas. Color changes as liquids are taken in, then returns to normal when neutralization is complete; helps identify the best time to handle with the least amount of risk. Instantly absorbs up to 98% concentrations of sulfuric acid.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty