Would you like to log in again?
Umbrella-Style Roof Leak Diverter Kit, PIG®
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TLS461-YW TLS461-CL TLS461-TR
10860-090EA 212.37 USD InStock
10860-090 10860-092 10860-094
Umbrella-Style Roof Leak Diverter Kit, PIG®
Spill Control Materials
All-in-one kit is perfect for quick responses to help prevent ruined inventory, damaged machinery and slip hazards.
  • Hassle-free setup in minutes
  • Hangs easily in problem spots
  • Resists punctures, UV and mildew for long life
Installs faster than any leak diverter in our offering; just hang the hook, pull off the cover and connect the hose for hassle-free setup in minutes. All-in-one kit is perfect for quick responses to help prevent ruined inventory, damaged machinery and slip hazards. Hangs easily in problem spots to catch nuisance leaks; flexible PVC drain hose channels them to a floor drain or collection container. Impermeable tarp is vinyl coated for strength and durability; resists punctures, UV and mildew for long life.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty