Would you like to log in again?
Plated Steel Drum Bung, PIG®, New Pig
Supplier: New Pig
DRM542
10860-608EA 40.41 USD InStock
10860-608
Plated Steel Drum Bung, PIG®, New Pig
Drum Storage
Plated Steel Drum Bung helps extend a drum's life.
  • Cut replacement costs
  • Plated steel drum bungs with Buna gaskets
  • Replaces damaged or lost bungs
Replacement parts to help you extend a drum's life and cut replacement costs. 3/4" plated steel drum bungs with Buna gaskets. Keep a box on hand to replace damaged or lost bungs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty