Would you like to log in again?
TALL TEST-TUBE CAP
Supplier: THERMO ELECTRON NORTH AMERICA LLC
Tall Test-Tube Cap^ Tall Test-Tube Cap, for GENESYS 30, Covers test-tubes up to 15cm/6 inches tall to allow measurements, while the sample compartment lid is openAllows for flexibility between samples, Storage Conditions: Ambient, Warranty: 2 year, Includes One cap to cover a tall test tube.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TALL TEST-TUBE CAP 10861-090 840-277600 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty