Would you like to log in again?
5-100MM PATH CYLINDRICAL CELL HOLDER
Supplier: THERMO ELECTRON NORTH AMERICA LLC
Cylindrical Cell Holder^ Cylindrical Cell Holder for GENESYS 30, 5-100mm Path, holds one cylindrical longpath cell 5 - 100mm pathlength, Allows flexibility with sampes and comes assembled in a removable sample compartment liner, Storage Conditions: Ambient, Warranty: 2 year
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
5-100MM PATH CYLINDRICAL CELL HOLDER 10861-094 840-277800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty