Would you like to log in again?
ACCESSORY KIT FOR HOSE ENTRY/EXIT
Supplier: THERMO ELECTRON NORTH AMERICA LLC
Accessory Kit For Hose Entry/Exit - G30^ Accessory Kit For Hose Entry/Exit - G30, Replacement sample compartment lid and liner with ports to allow hoses to enter and exit, Required to support to support thermostatted cell holder or flow cell, Storage Conditions: Ambient, Warranty: 2 year
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ACCESSORY KIT FOR HOSE ENTRY/EXIT 10861-098 840-278200 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty