Would you like to log in again?
10MM SQUARE CELL HOLDER
Supplier: THERMO ELECTRON NORTH AMERICA LLC
Cell Holder^ Cell Holder, Includes: 1 sample compartment liner with 10mm square cell holder Applications, Replaces the standard cell holder shipped with the instrument, Cuvette holder can be removed and replaced with this square cell holder, Storage Conditions: Ambient
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
10MM SQUARE CELL HOLDER 10861-110 840-278600 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty