Would you like to log in again?
Maintenance Spill Berm, Justrite®
Supplier: Justrite
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
28400 28418 28408 28416 28410 28406 28414 28422 28426 28402
10861-190EA 275.41 USD InStock
10861-190 10861-188 10861-200 10861-184 10861-174 10861-196 10861-176 10861-180 10861-192 10861-182
Maintenance Spill Berm, Justrite®
Spill Control Materials
Compact and flexible spill containment quickly contain minor spills and leaks during temporary storage of drums, machine parts, batteries, and equipment.
  • No assembly required; simply unfold and use
  • Encased urethane foam sidewalls offer easy roll-through access without the need for a ramp
  • Walls quickly spring back into place with no memory loss
Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner. Anchor stakes not included. Order optional grates to keep products and machinery above fluid.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty