Would you like to log in again?
Ground Protection Accessories, Justrite®
Supplier: Justrite
28336 28354 28338 28356 28360 28346 28352 28350 28342 28358 28334 28344 28362 28340 28348
10861-214EA 135.53 USD InStock
10861-214 10861-236 10861-226 10861-216 10861-238 10861-228 10861-232 10861-222 10861-234 10861-212 10861-224 10861-240 10861-230 10861-220 10861-218
Ground Protection Accessories, Justrite®
Spill Control Materials
Improve berm longevity without sacrificing functionality
  • Ground Mats - Create a smooth, uniform deployment area with this heavy-duty woven fabric. Place under berm for improved wear resistance against rocks and construction debris.
  • Track Mats - Position inside the berm for extra protection along tire paths. Thick durable, black woven fabric protects the floor and walls from tire abrasion caused by skidding, turning, and braking.
  • Track Runners - Place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion.
Use these berm accessories to increase the life and performance of your berm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty