Would you like to log in again?
Drip Pads, Justrite®
Supplier: Justrite
28461 28460 28459 28458
10861-266EA 150.07 USD InStock
10861-266 10861-268 10861-296 10861-270
Drip Pads, Justrite®
Spill Control Materials
Capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
  • Lightweight, flexible, and portable - great for containing small oil drips from leaky vehicles, industrial machinery equipment, hydraulic lines, pipes, and drums.
  • Made from UV- and chemical-resistant modified PVC.
  • Patented design feature includes a coated mesh outer layer to trap coarse debris.
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs. Helps comply with EPA for spill prevention (SPCC).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty