Would you like to log in again?
Water Filled Drain Cover, Justrite®
Supplier: Justrite
28470
10861-274EA 201.12 USD InStock
10861-274
Water Filled Drain Cover, Justrite®
Spill Control Materials
Water Filled Drain Covers seal drains before contaminants enter nearby water systems.
  • Preventative measure to seal drains
  • Easy-open, push plug
  • Resists damage caused by UV rays
Deploy cover as a preventative measure to seal drains before washdowns, maintenance, or other cleanup tasks that may allow contaminants into nearby water systems. Features easy-open push plug for filling with standard water hose. Durable EnGuard™V, Modified PVC coated fabric construction resists damage caused by UV rays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty