Would you like to log in again?
Pop-Up Pool, Justrite®
Supplier: Justrite
28319
10861-280EA 175.38 USD InStock
10861-280
Pop-Up Pool, Justrite®
Spill Control Materials
Durable, PVC-coated Pop-Up Pools contain spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery.
  • Folds tightly and stows easily
  • Embedded foam ring top raises containment wall as fluid level increases
  • Yellow color provides visibility and easy identification
  • Vinyl fabric, space-saving and lightweight
The durable PVC coated vinyl fabric space-saving and lightweight pools to contain spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty