Would you like to log in again?
Water-Filled Boom Diverter, Justrite®
Supplier: Justrite
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
28454 28450 28456 28452
10861-288EA 81.17 USD InStock
10861-288 10861-294 10861-290 10861-292
Water-Filled Boom Diverter, Justrite®
Spill Control Materials
Temporarily divert and control fluid for easy cleanup or recovery.
  • Helps comply with EPA for stormwater regulations
  • Redirects liquids away from work areas and storm drains
Use the water filled booms in storm water management, portable washdown/pressure washing diversion, spill diversion, and equipment and personnel decontamination. Flexible PVC film construction conforms to uneven surfaces such as asphalt or concrete to form a diversionary barrier away from storm drains or as a holding area for wet vacuum operations. Features easy-open push plug for filling with standard water hose.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty