Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Forced Air Circulation Incubators, Memmert

Supplier: Memmert

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

IF260PLUS230 IF160PLUS115 IF750230V IF55PLUS115 IF750115V IF260, 230 V IF160PLUS230 IF260, 115 V IF110, 230 V IF160, 230 V IF450PLUS115 IF55, 115 V IF30PLUS115 IF110PLUS230 IF160, 115 V IF30, 115 V IF750PLUS115 IF75, 115 V IF750PLUS230 IF110, 115 V IF110PLUS115 IF450115V IF75, 230 V IF450PLUS230 IF75PLUS115 IF30, 230 V IF55PLUS230 IF55, 230 V IF30PLUS230 IF75PLUS230 IF450230V IF260PLUS115
10861-324EA 10781.97 USD InStock
10861-324 10861-320 10861-322 10029-040 10028-928 10029-048 10028-920 10029-102 10029-046 10028-922 10029-100 10028-924 10029-044 10861-318 10028-926 10029-042 10861-456 10861-314 10861-316 10861-454 10861-312 10029-104 10029-050 10028-916 10028-918 10028-910 10028-912 10029-054 10029-098 10029-052 10029-096 10028-914
Forced Air Circulation Incubators, Memmert
Incubators Standard Incubators
The Generation 2012 range has natural and forced convection models with a temperature range up to +80 °C

SingleDISPLAY models; ideal for standard applications. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, programme time, time zones and daylight-saving time. One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log. Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiter TB acc. to DIN 12 880 which switches off the heating if the maximum permitted temperature is exceeded by approx. 10 °C, protecting the oven.

TwinDISPLAY models; ideal for complex ramping processes. Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters as above. Two PT100 sensors in a 4-wire circuit for mutual monitoring, taking over functions in case of an error. The HeatBALANCE function allows application specific adjustment of heating power distribution (balance) between the upper and lower heating areas. Logged protocol data is displayed on the ControlCOCKPIT (max 10,000 values correspond to approx. 1 week). Ethernet interface for reading out the protocol log and for uploading and implementing programmes and for online logging. Multiple overtemperature protection with audible and visual alarm: Electronic temperature monitoring TWW/TWB adjustable on display (independent adjustable temperature safety device class 3.1, which protects the samples and the oven) and mechanical temperature limiter TB (protection class 1) acc. to. DIN 12 880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual MIN / MAX values for over-/under-temperature.

There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application. All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. These incubators have an integrated data logger and digital timer which is adjustable from 1 minute to 99 days and 23 hours. All models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several e-mail addresses. INplus and IFplus units have a 4 hour sterilisation cycle at 160 °C. Complex ramping processes can be programmed quickly and easy for TwinDISPLAY models. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points. Stainless Steel color.

Certifications: CE Marked. UL listed.

Ordering information: Range of accessories and factory fitted options such as access ports, interior sockets, lighting and interior door are available on request. Please contact Avantor for further details prior to ordering as these can not be retro-fitted.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty