Would you like to log in again?
Peltier Cooled Incubators, IPP, Memmert
Supplier: Memmert
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IPP55115V IPP110PLUS230V IPP30115V IPP30PLUS115V IPP55PLUS115V
10861-468EA 7916.77 USD InStock
10861-468 10861-432 10861-460 10861-330 10861-328
Peltier Cooled Incubators, IPP, Memmert
Incubators Cooled Incubators
Peltier Cooled Incubator IPP* with Single DISPLAY
PELTIER COOLED INCUBATOR IPP Heating and cooling seamlessly with one system thanks to Peltier technology. In this respect, cooled incubators IPP not only contribute to climate protection, but it also achieves an additional decrease in operating costs of up 90% compared to compressor technology. This perfect development from the environmentally friendly and energy-saving heating/cooling technology by Memmert convinces by outstanding control precision and extremely small fluctuations.
Peltier Cooled Incubator IPPplus* with Twin DISPLAY. AtmoCONTROL Software.
Certifications: CE Marked.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty