Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Tissue Culture Dishes, Non-Treated, Sterilized, Non-Pyrogenic
Supplier: Avantor
TCD000070 TCD000060 TCD000090 TCD000100 TCD000150 TCD000035
10861-588CS 316.43 USD InStock
10861-588 10861-596 10861-586 10861-592 10861-594 10861-590
VWR® Tissue Culture Dishes, Non-Treated, Sterilized, Non-Pyrogenic
Culture Dishes Cell Culture Dishes
Cell and tissue culture dishes are ideal containers for cell growth and yields, and are also useful in sample separation, pre-treatment, and storage.
  • Available with 6 different diameters
  • Dish surface is smooth and free from striation
  • Easy and secure stacking
  • Sterilized by electron beam irradiation
  • Non-pyrogenic
Dishes are designed for use in applications where cell attachment is not desired. They offer an operating temperature range of −20 to +50 °C (−4 to +122 °F). Dishes are not autoclavable.
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
TC-treated: These dishes are vacuum-gas plasma or corona treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture dishes are ideal for applications where cell attachment is not desired.
Certifications: ISO 13485 2003 and ISO 9001 2008 certified.
Packaging: Packaged in durable zip resealable self-standing plastic bags that allow dishes to remain upright and reduce contamination. Every bag is labeled with Lot Number for quality traceability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty