Would you like to log in again?
VWR® Cell Culture Flasks Treated for Increased Cell Attachment, Sterile
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CAF011850 CAF011600 CAF011250 CAF012850 CAF011025 CAF012250 CAF011050 CAF012050 CAF012025 CAF012600
10861-654CS 326.06 USD InStock
10861-654 10861-644 10861-646 10861-672 10861-650 10861-640 10861-652 10861-642 10861-670 10861-648
VWR® Cell Culture Flasks Treated for Increased Cell Attachment, Sterile
Flasks Cell Culture Flasks
Increased cell attachment surface treatment can improve cell spreading and attachment.
  • Available with 6 different growth surface areas
  • Increased Cell Attachment surface treated
  • Uniform well volume ensures an equal growth surface area
  • Flat well bottom and round bottom plates available
  • Well surface is smooth and free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on the lid to reduce evaporation
  • Single position lid reduces the risk of cross-contamination and handling mistakes
  • Wells are labeled with alphanumeric code for easy identification
  • Suitable for all common instruments and automation
  • Sterilized by electron beam irradiation
  • DNase- and RNase-free, and non-pyrogenic
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
TC-treated: These dishes are vacuum-gas plasma or corona treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture dishes are ideal for applications where cell attachment is not desired.
Certifications: ISO 13485 2003 and ISO 9001 2008 certified.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty