Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Cell Culture Dishes Treated for Increased Cell Attachment, Sterile
Supplier: Avantor
CAD010035 CAD010100 CAD010150 CAD010090 CAD010060 CAD010070
10861-656CS 299.46 USD InStock
10861-656 10861-662 10861-680 10861-682 10861-660 10861-658
VWR® Cell Culture Dishes Treated for Increased Cell Attachment, Sterile
Culture Dishes Cell Culture Dishes
Increased cell attachment surface treatment can improve cell spreading and attachment.
  • Available with 6 different diameters
  • Increased cell attachment surface treated
  • Flat bottom, uniform wall thickness ensures distortion-free bottom
  • Rim on the bottom side of the lid mates with the dish brim for easy and secure stacking
  • Numeric scale divides dish into quadrants for easy orientation
  • DNase- and RNase-free, and non-pyrogenic
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
TC-treated: These dishes are vacuum-gas plasma or corona treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture dishes are ideal for applications where cell attachment is not desired.
Certifications: ISO 13485 2003 and ISO 9001 2008 certified.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty