Would you like to log in again?
Multi-Layer Foil Gas Sampling Bags, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22968 22967 22951 22966 22952 22950 22953
10862-990PK 220.34 USD InStock
10862-990 10862-578 10862-986 10862-576 10862-984 10862-988 10863-020
Multi-Layer Foil Gas Sampling Bags, Restek
Bags Sample Bags
Sampling bags are a low-cost, whole-air sampling device for high-level VOCs and permanent gases.
  • Good stability for low molecular weight compounds
  • Chemically inert with light and moisture protection
  • Not recommended for low ppm VOCs due to background levels
  • Protective five layer barrier minimizes gas permeability
  • 60 gauge nylon (outer layer)
  • Metalized aluminum/Polyethylene
  • 0.0003" aluminum foil/0.002" polyethylene (inner layer)
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplace atmospheres, ambient, indoor air quality, and breath analysis. Choose the film type appropriate for your application. All our bags feature a polypropylene combo valve with hose connection to fit 3/16" Int.Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty