Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ALTEF Gas Sampling Bags, Restek

Supplier: Restek
26329 26331 22958 22962 26330 22960 26328 22963 22961
10862-684PK 256.52 USD InStock
10862-684 10863-044 10863-046 10862-710 10862-730 10862-702 10862-708 10862-706 10862-728
ALTEF Gas Sampling Bags, Restek
Bags Gas Bags
ALTEF Gas Sampling Bags are an excellent low-cost alternative to Tedlar bags.

  • For collection of most VOCs
  • Very low VOC and sulfur background
  • Not recommended for ketones, acetates, hydrogen sulfide, or permanent gases
  • Durable PVDF film
  • No additives, fillers, or pigments

Sampling bags are a low-cost whole air sampling device for high level VOCs and permanent gases.

All bags feature a polypropylene combo valve with hose connection to fit ³/₁₆" Int.Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.

Durable PVDF film is resistant to abrasion and chemicals, including most acids and organic compounds.

Thickness: 0.003"
Operating Temperature: –22 to 150 °C
Tensilse Strength: 6100 ps
Specific Gravity: 1.78 g/ml
Oxygen Permeability: 58 cc/m² × day
Water Vapor Permeability: 12 to 15 g/m² × day
Carbon Dioxide Permeability: 172 cc/m² × day

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty