Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Small Cylinder Stand, Restek

Supplier: Restek
24129
10862-714EA 138.7 USD InStock
10862-714
Small Cylinder Stand, Restek
Support Stands
This cylinder stand is designed to support small-diameter cylinders, such as 104 L and 110 L disposable cylinders.

  • Supports and stabilizes 8 cm diameter disposable gas cylinders
  • Fits cylinders up to 3 3/8" (8.57 cm) in diameter
  • Adjustable screw secures cylinder in place
  • Base diameter: 7.75" (19.68 cm)

It is a simple, safe, and economical way to stabilize the position of small cylinders, while keeping them within close proximity. The stand is constructed of heavyweight painted steel and includes an adjustable screw for safely securing cylinders.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty