Would you like to log in again?
Passive Air Sampling Kits-Integrated (Stainless Steel and Siltek®-Treated), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
24161 22100 24216 22101 24169 24168 24166 24160 24167 24165
10863-098EA 1180.98 USD InStock
10863-098 10862-754 10862-616 10862-966 10863-060 10862-614 10863-062 10862-738 10862-580 10863-064
Passive Air Sampling Kits-Integrated (Stainless Steel and Siltek®-Treated), Restek
Samplers Air Samplers
Restek’s passive air sampling kit incorporates all the hardware necessary to collect air samples and is easy to assemble for field sampling.
  • Provide accurate integrated sampling without a sampling pump
  • Siltek®-treated components ensure accurate sampling of active components
  • Excellent for sampling times from 0.5 hour to 125 hours
The improved filter design greatly reduces the number of potential leak sites.The passive air sampling kit is available in seven sampling flow ranges and in stainless steel or Siltek®-treated finish. The stainless steel kit is ideal to partner with the Restek® TO-Can® air sampling canister for TO-14A and TO-15 methods. Use the Siltek®-treated version with the Restek® SilcoCan® air sampling canister when collecting low-level volatile sulfur compounds or other active compounds.
Accessories information: Vacuum/pressure gauge included in kit; air sampling canisters sold separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty