Would you like to log in again?
Replacement Combination Vacuum/Pressure Gauges, Restek
Supplier: Restek
24104 24100 24108
10862-788EA 183.67 USD InStock
10862-788 10862-790 10862-818
Replacement Combination Vacuum/Pressure Gauges, Restek
Manometers
2-inch vacuum/pressure gauges, 316 stainless steel with 1/8" NPT fitting and center back mount.
  • Combination gauges
  • For use with canisters
Recommended for use with canisters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty