Would you like to log in again?
Canister Air Sampling Timer, Restek
Supplier: Restek
24267
10862-858EA 3094.25 USD InStock
10862-858
Canister Air Sampling Timer, Restek
Timers Digital Timers
These timers are designed to simplify both automated and manual air sampling.
  • Program up to 12 timed events
  • Capable of both manual and automated operation
  • Perfect for either grab or time-integrated sampling
  • Long battery life
  • Recharges conveniently using the USB port on any PC
The easy-to-use keypad and graphic display facilitate the programming of up to 12 timed events.
Features include solenoid valve for sampling control, ¹/₄" inlet and outlet fittings, highly inert stainless steel flow path, and waterproof exterior for outdoor use.
They offer the convenience of remote start/stop sampling and permit intermittent sampling throughout a test period. The LCD remains in sleep mode when not in use, greatly extending battery life. Timers are compatible with any canister and flow controller.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty