Would you like to log in again?
Dual Canister Sampling Manifold (Stainless Steel & Siltek®-Treated), Restek
Supplier: Restek
24998 24999
10862-940EA 378.79 USD InStock
10862-940 10862-924
Dual Canister Sampling Manifold (Stainless Steel & Siltek®-Treated), Restek
Manifolds
Field duplicates of canister samples frequently result in analyte concentrations with high relative standard deviations.
  • Duplicate sampling with all canister sizes using one or two flow controllers
  • Precise dimensions (9.5" wide x 3.5" high)
  • Provides accurate splitting of sample between two canisters
  • One-piece design means fewer leaks
  • Thick-walled stainless steel tubing is rugged enough for field use
In addition, field duplicates do not differentiate laboratory performance from sampling variability. Restek’s dual canister sampling manifold (DCSM) minimizes sampling variability through a single sample inlet and flow controller by which the sample is evenly collected between two canisters. Use of a single flow controller eliminates flow rate variability, as well as environmental variables common with collocated samples. The DCSM may also be used with two flow controllers to monitor individual canister vacuum.
Ordering information: Do not use the DCSM as a handle to pick up 2 canisters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty