Would you like to log in again?
Veriflo® Flow Controllers (Stainless Steel and Siltek® Treated), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
24239 24261 24229 24238 24260 22102 24255 24263 24262 22103 24264
10862-992EA 1127.63 USD InStock
10862-992 10862-586 10863-036 10863-234 10862-996 10862-700 10862-994 10863-070 10862-712 10862-998 10863-192
Veriflo® Flow Controllers (Stainless Steel and Siltek® Treated), Restek
Flow Controllers
Veriflo® 423XL flow controllers are offered in a Siltek®-treated and stainless steel version.
  • Treated and non-treated version
  • Available with or without critical orifice
The flow device is available with or without a critical orifice. The critical orifice in a Veriflo® flow controller is interchangeable.
Accessories information: Order orifices for alternate sampling times, or replacement orifices, separately.
Ordering information: Vacuum gauge sold separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty