Would you like to log in again?
SilcoCan® Air Sampling Canisters with RAVE™ Valve, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
27400 27415 27407 22092 27408 22091 27412 27404 27405 22093 27411 27403 27409 27413 27401 22090
10863-142EA 665 USD InStock
10863-142 10863-188 10862-736 10862-824 10862-726 10862-734 10862-724 10862-834 10862-826 10863-140 10862-860 10862-864 10862-854 10862-832 10862-862 10862-554
SilcoCan® Air Sampling Canisters with RAVE™ Valve, Restek
Samplers Air Samplers
Ideal for low-level reactive sulfur (5–20 ppb), TO-14A, or TO-15 compounds.
  • For low-level compounds
  • Prevents sample adsorption
  • Excellent storage stability
  • High-quality seal
For ultimate inertness, SilcoCan® air sampling canisters feature our unique Siltek® treatment technology
Whether you are sampling for TO-14A, TO-15, or reactive sulfur compounds, SilcoCan® canisters are your best choice for inertness. In Tedlar® bags, the stability of low-level (100 ppbv) sulfur volatile organic compounds (VOCs) is poor, even within 24 hours of sampling. Sulfur compounds react with the metal surface in electropolished canisters, so they are unsuitable for collecting and storing low-level sulfur VOCs. SilcoCan® air sampling canisters, which feature a Siltek®-treated surface, offer excellent storage stability for sulfur VOCs at very low levels (5–20 ppbv), under dry or humid conditions. The versatility of the SilcoCan® canister makes it an excellent choice for collecting and storing TO-14A or TO-15 compounds.
Canisters and valves made of 304 and 316 stainless steel to withstand the rigors of field work. Both 2-port and 3-port valves available; 3-port valve includes -30" Hg/60 psi vacuum/pressure gauge (other gauges available).
Canisters and valves made of 304 and 316 stainless steel to withstand the rigors of field work. Both 2-port and 3-port valves available; 3-port valve includes -30" Hg/60 psi vacuum/pressure gauge (other gauges available).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty