Would you like to log in again?
Ashcroft® Test Gauges, Restek
Supplier: Restek
24268 24285
10862-880EA 1010.25 USD InStock
10862-880 10863-206
Ashcroft® Test Gauges, Restek
Valves Vacuum Valves
High-accuracy test gauges are recommended for verifying the vacuum/pressure in canisters before and after sampling.
  • Accurate measurement of vacuum to –30" Hg and pressure to 60 psi
  • Available in both analog and digital formats
  • Accuracy to +/– 0.25%
  • Gauge connector to canister valve available
The 6-inch face on the analog gauge allows for easy reading. The digital gauge operates on two AAA batteries and offers an unambiguous readout. Both gauges have an accuracy of +/– 0.25% and all-metal wetted parts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty