Would you like to log in again?
Magid® TouchMaster® Inspection Gloves, Magid
Supplier: Magid
661J 660 661
11000-294PK 47.19 USD InStock
11000-294 11000-265 32889-460
Magid® TouchMaster® Inspection Gloves, Magid
Gloves
Medium weight cotton lisle inspection gloves are 100% cotton two-piece lisle inspection glove constructed of cool and breathable bleached white cotton.
  • Form-fitting and cut and sewn for the most comfortable fit
  • Color-coded sizes for easy sorting
They can be worn alone to protect delicate products from smudges, finger prints or skin oils, or used as a liner to absorb moisture. They are an excellent choice for jobs in the plastics industry.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty