Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crews® Protective Goggles, MCR Safety

Supplier: MCR Safety

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Crews®
2225R 2220R 2230R 2220 2235R 2237R
11000-540CS 704.28 USD InStock
11000-540 11000-398 80081-784 80081-782 80081-776 80081-778
Crews® Protective Goggles, MCR Safety
Goggles
Goggles provide a wide, unobstructed field of vision and are available in a variety of styles to protect against impact and splash.

  • Wide, unobstructed field of vision
  • Can be worn with most half-mask respirators and will fit comfortably over most prescription eyewear
  • Transparent PVC frame and clear 0.06mm thick polycarbonate lens with scratch-resistant Duramass® coating
  • Latex-free rubber straps provide secure fit on all styles
  • Lenses provide 99.9% UV protection
  • Goggles meet or exceed ANSI Z87.1+ safety standards

All goggles feature a transparent PVC frame and clear 0.06mm thick polycarbonate lens with scratch-resistant Duramass® coating. Latex-free rubber straps provide a secure fit on all styles. Lenses provide 99.9% UV protection.

They can be worn with most half-mask respirators and will fit comfortably over most prescription eyewear.

Certifications: Goggles meet or exceed ANSI Z87.1+ safety standards.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty