Would you like to log in again?
Silver Shield®/4H® Booties, Honeywell Safety
Supplier: Honeywell Safety
Silver shield® 4H® Silver Shield® 4H®
SSB SSB
11000-628CS 1301.02 USD InStock
CA11000-629 11000-628
Silver Shield®/4H® Booties, Honeywell Safety
Disposable Garments
Booties feature a five-layer fabric compacted into 2.7 mil
  • Five-layer fabric compacted into 2.7 mil
  • High-energy polymer (EVOH) with a low-energy polymer (PE), impervious to at least 90% of all organic chemicals for 4 hours at 35 °C (95 °F)
  • Each layer acts as a barrier against various epoxies and industrial chemicals
  • Wear as a liner or as a cover
  • One size fits all
Laminating a high energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35 °C (95 °F). Each layer acts as a barrier against various epoxies and industrial chemicals. Can be worn as a liner under regular boots, or as a cover over shoes or boots. One size fits all.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty