Would you like to log in again?
Silver Shield®/4H® Coat Aprons, Honeywell Safety
Supplier: Honeywell Safety
Silver Shield® 4H®
SSCA/XL SSCA/XL SSCA/S SSCA/L SSCA/L SSCA/M SSCA/M SSCA/S
11000-632CS 1126.45 USD InStock
CA11000-638 11000-632 CA11000-632 11000-634 CA11000-634 11000-636 11000-638 CA11000-636
Silver Shield®/4H® Coat Aprons, Honeywell Safety
Disposable Garments
Aprons feature a five-layer fabric compacted into 2.7 mil.
  • Each layer acts as a barrier against various epoxies and industrial chemicals
  • Impervious to at least 90% of all organic chemicals for 4 hours at 35 °C (95 °F)
  • Cut from a single piece of film to prevent penetration of liquid chemicals
  • Sleeves with flexible wrist openings, buckle closure attached to neckband, two waistbands that tie in the back
Each apron weighs less than 198 g (7oz).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty