Would you like to log in again?
EYEWEAR PROTECTV NORTON180 CLR
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Norton 180* Protective Eyewear^ Made of lightweight 100% polycarbonate. Clear, scratch-resistant one-piece wraparound lens. Vented sideshield temples reduce heat buildup and lens fogging. Frosted brow reduces glare. Four-post hinge system increases impact resistance. Meet ANSI Z87.1-1989 standard.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EYEWEAR PROTECTV NORTON180 CLR 11000-926 T18000 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty