Would you like to log in again?
Autokit 3-HB R2 Set, Fujifilm Wako Chemicals USA
Supplier: Fujifilm Wako Chemicals USA
413-73601 413-73601
11001-662EA 1188.31 USD InStock
CA11001-662 11001-662
Autokit 3-HB R2 Set, Fujifilm Wako Chemicals USA
Assays Protein Assays
The Autokit 3-Hb R2 Set is an in vitro assay for the quantitative determination of 3-hydroxybutyrate (3-HB) in serum or plasma.
  • Utilizes cyclic enzymatic reactions
  • High sensitivity and high specificity
  • Quantitative determination of 3-HB in serum or plasma
The Autokit 3-Hb R2 Set is an in vitro assay for the quantitative determination of 3-hydroxybutyrate (3-HB) in serum or plasma. It utilizes cyclic enzymatic reactions for high sensitivity and high specificity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty