Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gifu Anaerobic Broth, HiMedia

Supplier: HiMedia Laboratories
M1801-500G M1801-500G
11007-214EA 227.27 USD InStock
CA11007-214 11007-214
Gifu Anaerobic Broth, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Gifu Anaerobic Medium (GAM Broth) is recommended as a general culture medium for cultivation and isolation of anaerobic bacteria and to test their susceptibility to antibiotics other than sulpha drugs.

  • Light yellow to brownish yellow homogeneous free flowing powder
  • Forms amber coloured clear solution in tube
  • Reaction of 5.9% w/v aqueous solution at 25 °C, pH: 7.3±0.1

Composition of the medium is as follows: Peptic digest of animal tissue, papaic digest of soyabean meal, protease peptone, digested serum, yeast extract, beef extract, liver extract, dextrose, potassium dihydrogen phosphate, sodium chloride, starch, L-cysteine hydrochloride, sodium thioglycollate.

GAM broth is to be storted below 30 °C and the prepared medium at 2 - 8 °C.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty