Would you like to log in again?
Whatman™ PZ 001 Tweezers
Supplier: Whatman products (Cytiva)
Whatman™
10477602 10477602
11008-548EA 65.96 USD InStock
11008-548 CA11008-548
Whatman™ PZ 001 Tweezers
Forceps
The stainless steel forceps with smooth angled jaws (104 mm long) are highly suitable for handling membrane filters
They are autoclavable and can be flame-sterilized with ethanol.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty