Would you like to log in again?
Whatman™ Grade 1573 and 1573 ½ Qualitative Filter Papers, Wet Strengthened Grades, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10314765 10314753 10314726 10314744 10314752 10314766 10314747 10314726 10314714 10314766 10314751 10314712 10314765 10314753 10314714 10314712 10314744 10314747 10314752 10314745 10314751 10314745
10034-734PK 318.97 USD InStock
CA11012-790 CA11012-770 10034-734 CA14224-412 CA11012-786 14224-412 10034-724 CA11012-788 76423-756 10034-730 CA76423-756 CA14224-416 11008-574 10034-732 CA11008-574 CA76423-754 CA11012-778 14224-416 76423-754 CA11012-826 10034-726 10034-728
Whatman™ Grade 1573 and 1573 ½ Qualitative Filter Papers, Wet Strengthened Grades, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A fast filter paper with high wet strength. It has a very smooth surface making it easy to scrape or wash off precipitate.
  • Folded filters for easy use in applications involving a conical filter funnel
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty