Would you like to log in again?
Ore Sample Bags, Contract Sewing & Manufacturing
Supplier: Contract Sewing & Manufacturing
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MICRO13X20 AC0912CDTL AC0912CDTL 105280 068-020-01
75796-892CS 1790.75 USD InStock
75796-892 80700-192 11017-678 CA11017-678 80701-644
Ore Sample Bags, Contract Sewing & Manufacturing
Bags Sample Bags
Sample bags available in a variety of materials.
  • Customizable
  • Accord bags are economical and strong
Micro Por Plus PE bags provide the right combination of properties for the most accurate assay results. Material will handle up to 232 °C (450 °F). Bags are extremely strong and rugged and are inert to most organic chemicals (acids, base, salts). Available drawstring and 5×10 cm (2×4") durable, waterproof tag. Material strength holds up very well over time.
Other materials (Cotton or Canvas), custom sizes and additional options (such as bar-coding) are available on a custom quote basis. Contact your Avantor sales representative for more information.
Spun-bonded polypropylene fabric drains quickly, while retaining fines. Great for collecting large, wet samples. Fibers are evenly distributed to allow for some stretch.
Ordering information: Accord material will weaken and break down under prolonged UV (outdoor sunlight) exposure. Recommend covering with tarp (outdoor storage) or storing indoors.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty