Would you like to log in again?
LAMP HCL 1.5 ALUMINIUM 0 0 0 0 0 0 0
Supplier: PHOTRON PTY LTD.
Hollow Cathode Lamp Aluminium^ Hollow Cathode Lamp, Material: 1.5 Aluminium, Window material: Quartz, To suit all Atomic Absorption Spectroscopy Instruments, Two Pin Standard, Primary Wavelength: 396.2nm, Alternative Wavelengths: 308.2, Approx 309.3
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAMP HCL 1.5 ALUMINIUM 0 0 0 0 0 0 0 11017-768 P801 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty