Would you like to log in again?
Hollow Cathode Lamps to Suit All AAS Systems, Photron
Supplier: Photron
P833 P814 P820 P831 P833 P821 P873P873 P849 P813 P813 P821C P812 P852 P821C P851 P804 P839 P808 P806 P809 P527 P802 P851C P828 P867 P826 P851 P832 P840 P851C P803C P836 P872 P871 P834 P524 P870 P821 P872 P836 P814 P855 P834 P803 P803C P832 P524 P828 P527 P803 P808 P867 P841
11018-528EA 802.08 USD InStock
11018-528 CA11017-770 CA11017-772 CA11017-774 CA11017-972 CA11017-806 11017-838 CA11017-808 11017-834 11017-832 CA11017-844 CA11017-846 CA11017-968 CA11017-848 CA11018-528 CA11017-782 CA11017-980 CA11017-784 CA11017-982 CA11018-522 11017-972 11017-772 11017-968 11018-522 CA11017-778 CA11017-978 11017-808 CA11017-790 CA11017-792 CA11017-870 CA11017-794 CA11017-992 11017-982 11017-782 CA11017-828 CA11017-908 CA11017-984 CA11017-864 CA11017-822 CA11017-868 11017-818 11017-830 11017-794 11017-992 11017-792 CA11017-838 11017-868 11017-822 CA11017-830 CA11017-832 CA11017-876 CA11017-834 11017-908
Hollow Cathode Lamps to Suit All AAS Systems, Photron
Lights Hollow Cathode Lamps
Hollow cathode lamps produced by Photron are designed and manufactured to achieve all of the following fundamental requirements for a spectral line source: Intense emission of resonance (ground state derived) lines. Narrow line width, for maximum sensitivity and linearity. Minimal spectral interference from continuum emission, present in the cathode. Rapid warm up to produce stable long term light emission. Noise free operation. Long running life and a 5 year shelf life. Cathode materials and geometry achieve a combination of spectral purity and a suitable sputtering rate. (A high sputtering rate will give high intensity at the expense of lamp life, sensitivity and linearity).
Photron's P800 (37mm or 1.5” Dia.) series is designed for use in AAS systems manufactured by GBC Scientific, Varian, Analytic Jena, Shimadzu, and other makes
Photron's P500 Multi Element (37mm or 1.5” Dia.) series is designed for direct use in AAS systems manufactured by GBC Scientific, Varian, Analytic Jena, Shimadzu, Hitachi, Thermo Jarrel Ash, Unicam, and other makes.
SMITH-HIEFTJE background correction. Photron lamps have been tested and found to work as well as lamps sold specifically for this technique, however due to the high energy pulse used, some elements will be consumed at a higher rate reducing lamp life.
Ordering information: Contact your Avantor sales representative for more information. Wavelength listed is the primary wavelength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty