Would you like to log in again?
Bead Pliers, Grobet USA
Supplier: Grobet USA
46.329 46.329
11019-240EA 65.72 USD InStock
11019-240 CA11019-240
Bead Pliers, Grobet USA
Pliers
These bead pliers are specially designed with a bent tip for picking up beads
  • Constructed from hardened steel for dependable, long-lasting use
  • Spring-loaded handle ensures ease of use
Curved tips taper to 1/16" for setting small stones. Box joint for alignment with built-in spring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty