Would you like to log in again?
A.P. Green Fire Assay Crucibles, HarbisonWalker
Supplier: Harbison-Walker
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
54691 54694 54684 112531 54694 54688 83472 54697 54686 54697 54698 54686 54684 54688 54691
80704-052CS 111.48 USD InStock
80704-052 11019-260 80704-054 11019-264 CA11019-264 11019-262 CA11019-266 11019-256 CA11019-260 11019-266 CA11019-262 11019-258 CA11019-256 CA11019-246 CA11019-258
A.P. Green Fire Assay Crucibles, HarbisonWalker
Crucibles
A. P. Green Fire Assay Crucibles have several features which make them ideal for mining assays.
  • Withstand thermal shock
  • Allow for more heats per crucible
  • Multiple size options available
Their scientific mix of material batches, proper pressing with accurate dyes, and the correct graduating temperatures at which they are burned, provide the highest degree of mechanical strength to withstand severe thermal shock. The high quality materials used in their manufacture, coupled with carefully managed production, ensures more heats per crucible, thus providing more value. Combine all these features with the convenience of several available sizes and shapes, and A.P. Green Fire Assay Crucibles become the perfect choice for mining applications.
Ordering information: Multiple size options are available. Contact your Avantor sales Representative for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty