Would you like to log in again?
Oakton pHTestr 10 Waterproof BNC Pocket pH Tester, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35634-14
11026-016EA 262.21 USD InStock
11026-016
Oakton pHTestr 10 Waterproof BNC Pocket pH Tester, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Customizable to your application – use with any pH electrode with a BNC connector.
  • Versatility of a handheld meter at a pocket price
  • IP67-rated protects meter from water, dust, and shock while offering chemical resistance
  • Meter floats, accidental drops into water will not result in lost or damaged meters
Ordering information: Includes protective plastic storage case, lanyard, and batteries.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty