Would you like to log in again?
Lens Cleaning Solution, C-Clear
Supplier: C-Clear
21 26 44
89094-546CS 247.05 USD InStock
89094-546 11200-000 11200-002
Lens Cleaning Solution, C-Clear
Cleaning Agents Lens Cleaning Solutions
C-Clear lens cleaning solution is specially designed for the industrial setting.
  • Contains no silicone
  • Effectively cleans glass, plastic and polycarbonate lenses
  • Used on sideshields, face shields, goggles, respirators and CRT screens
This environmentally safe fluid cuts through grease and oil while leaving an anti-fog/anti-static coating on the lenses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty