Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Supplier: Cardinal
Convertors®
92510 92713 92309 90422 90308 92923 90408 90713 91822 90610 90411 92318 90515 90810 92318 92309 92308 91616 91015 91218 92713 90411 90713 92152 92322 90408 92515 91315 91015 90810 92510 90515 92322 92515 90610 91315 90422 92923 91822 90308 91616 91218 92152 92308
11213-036CS 433.82 USD InStock
CACV92309 CA11213-239 CA11213-245 CA11213-025 11213-036 CA11213-027 11213-035 11213-239 CA11213-021 CA11213-044 CA11213-243 CA11213-023 11213-237 11213-236 11213-038 CA11213-062 CA70000-414 11213-033 CA70000-418 11213-230 11213-031 CA70000-416 CA11206-410 CV-92309 CA11206-415 CA11206-414 CA89045-093 11213-245 11213-025 CA11213-236 11213-023 CA70000-409 11213-243 CA11213-230 CA11213-033 11213-029 CA11213-035 11213-027 11213-040 11213-062 11213-044 11213-241 11213-021 11213-042
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Bags Sterilization Pouches
Peel-open pouches are designed for quick, easy presentation of sterile items.
  • Can be used in steam or EO sterilization
  • Chemical process indicators on every pouch identify the sterilization process used
Heat-seal pouches can be heat-sealed or taped shut. Self-seal pouches feature a pressure-sensitive closure that is prefolded to help ensure the integrity of the seal.
Items within the pouch are easily viewed through the blue-tinted 2 mil-thick film side. The other side is made of tough, moisture-resistant, surgical-grade paper, which provides high porosity for fast sterilization while maintaining an effective bacterial barrier.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty