Would you like to log in again?
Kapak SealPAK 400 and 500 Series Pouches, Ampac Flexibles
Supplier: Ampac Flexibles
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Kapak®
404B-2M 504B-1M 400B-2M 400-24 500-24 402-24 508-24 406-24 502-24 503-24 502B-1M 506B-500 403-24 505B-1M 408-24 508B-500 506-24 404-24 505-24 405B-2M 504-24 402B-2M 405-24 500B-1M 503B-1M
11214-495CS 3035.07 USD InStock
11214-495 11214-550 11214-334 11214-390 11214-491 11214-394 82017-786 10027-534 10027-576 11214-527 11214-562 82007-178 11214-564 82007-176 11214-301 11214-367 11214-469 10027-536 11213-918 11213-916 11214-560 10027-540 82007-182 11214-338 82007-180
Kapak SealPAK 400 and 500 Series Pouches, Ampac Flexibles
Bags Sample Bags
Boilable, freezable, and microwavable polyester/polyethylene (PET/PE) laminated Kapak pouches combine tensile strength with strong heat seals and superior barrier properties.
Regular weight 2.5 mil material is best for moderate protection from handling and chemical or environmental contact. Tear notches included. Heavy-duty 4.5 mil material is best for institutional and industrial use; they are also suitable for long-term storage and mailing. Tear notches not included.
These pouches will sustain temperatures from –56 to +115 °C (–70 to +240 °F). Safe for use with concentrations of formaldehyde, most acids, alkalies, greases, oils, and organic solvents. Transparent, airtight, moisture, and odor-proof. They can withstand infrared and gamma rays. These pouches can be surface sterilized using VHP. Ideal for histology and pathology specimens, as well as safe biological waste disposal in medical, institutional, and industrial facilities. Seal using vacuum or commercial heat-sealing equipment
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty