Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Terminal® Biohazard Benchtop Keeper™ Containers, Whitney Products
Supplier: Whitney Medical Solutions
Terminal® Biohazard Benchtop Keeper™ Terminal® Keeper™
BH2003 BH2002
11214-711CS 331.28 USD InStock
11214-711 11214-708
Terminal® Biohazard Benchtop Keeper™ Containers, Whitney Products
Boxes Waste Disposal Boxes
Use large or small containers for the collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides.
  • For collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides
  • Sturdy, plastic-lined paperboard construction
  • Leak-resistant and autoclavable
Constructed of sturdy, plastic-lined paperboard. Disposable containers are leak-resistant and autoclavable. Self-locking top for safe closure.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty