Would you like to log in again?
SP Bel-Art Utility Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
H13176-0812 H13178-0046 H13178-0612 H13178-1018 H13178-1218 H13178-0810 H13178-0612 H13178-0068 H13176-1220 H13176-1012 H13176-0818 H13178-0812 H13176-0612 H13176-1016 H13178-1012 H13178-1214 H13178-1220 H13178-1016 H13178-1216 H13176-1014 H13178-0816 H13176-1218 H13178-0610 H13176-0816
74950-058CS 634.98 USD InStock
74950-058 11215-359 11215-348 13197-294 13197-304 13197-314 11215-392 47750-768 13197-322 11215-280 11215-390 13197-298 11215-259 11215-202 11215-257 11215-235 74950-406 11215-246 11215-388 13197-308 47748-162 47748-164 13197-306 13197-316
SP Bel-Art Utility Bags, Bel-Art Products, a part of SP
Bags Storage Bags
These all-purpose polyethylene bags are available in two weights.
  • Polyethylene construction
  • Latex-free
  • Not autoclavable
  • Reusable and transparent
They are ideal for shipping or storage and are especially useful as sample bags.
Certifications: FDA approved material.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty